missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMF1 Recombinant Rabbit Monoclonal Antibody (24GB1965)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen TMF1
Clone 24GB1965
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275798
GREENER_CHOICE
View Documents
Invitrogen™
MA557766
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274877
View Documents
Invitrogen™
MA557767
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TMF1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
Specifications

Specifications

TMF1
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
TMF1
7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1
TMF1
Primary
Affinity Chromatography
24GB1965
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P82094
7110
A synthesized peptide derived from human TMF (aa 1000-1093).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.