missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19728-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to TMF1.
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.Specifications
| TMF1 | |
| Polyclonal | |
| Unconjugated | |
| TMF1 | |
| TMF1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P82094 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7110 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction