missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531184
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG, mouse testis. Predicted reactivity: Mouse (86%), Rat (83%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
Specifications
| TMF1 | |
| Polyclonal | |
| Unconjugated | |
| TMF1 | |
| 7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114206, 232286, 7110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| B9EKI3, P82094 | |
| TMF1 | |
| Recombinant protein encompassing a sequence within the center region of human TMF1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction