missing translation for 'onlineSavingsMsg'
Learn More

TMEPAI Rabbit anti-Human, Clone: PMEPA1/6421R, Novus Biologicals™

Product Code. 18375772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375772 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375772 Supplier Novus Biologicals Supplier No. NBP30724620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEPAI
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PMEPA1/6421R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein
Host Species Rabbit
Immunogen Recombinant full-length human TMEPAI protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56937
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.