missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEPAI, Mouse anti-Human, Clone: PMEPA1/2698, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79758-100UG
This item is not returnable.
View return policy
Description
TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| TMEPAI | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| PMEPA1/2698 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| PMEPA1 | |
| Recombinant full-length human TMEPAI protein | |
| 100 μg | |
| Signal Transduction | |
| 56937 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction