missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEPAI Antibody (PMEPA1/2697), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79892AF405
This item is not returnable.
View return policy
Description
TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TMEPAI | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PMEPA1 | |
| Recombinant full-length human TMEPAI protein (Uniprot: Q96W9) | |
| 0.1 mL | |
| Signal Transduction | |
| Human | |
| IgG κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PMEPA1/2697 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction