missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM98 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14731-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a transmembrane protein. A missense mutation in this gene result in ophthalmos 4 (NNO4). Alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
| TMEM98 | |
| Polyclonal | |
| Unconjugated | |
| TMEM98 | |
| Protein TADA1, TMEM98, transmembrane protein 98 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103743, 26022 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q91X86, Q9Y2Y6 | |
| Tmem98 | |
| TMEM98 Fusion Protein Ag6492 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction