missing translation for 'onlineSavingsMsg'
Learn More

TMEM97 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18407924
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407924 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407924 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP130437JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM97 Polyclonal antibody specifically detects TMEM97 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM97
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias MAC30Protein MAC30, transmembrane protein 97
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse TMEM97 protein (within residues 20-100. [Swiss-Prot# Q8VD00]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Cytokine Research, Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 27346
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.