missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TMEM9 Recombinant Protein Antigen
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1M Urea, pH 7.4. |
| Gene ID (Entrez) | 252839 |
| Molecular Weight (g/mol) | M.W. Theoretical: 23 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | Dermal papilla-derived protein 4, DERP4, TMEM9A, transmembrane protein 9 |
| Gene Symbol | TMEM9 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | TMEM9 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?