missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM82 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | TMEM82 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TMEM82 Polyclonal specifically detects TMEM82 in Human samples. It is validated for Western Blot.Specifications
| TMEM82 | |
| Polyclonal | |
| Rabbit | |
| A0PJX8 | |
| 388595 | |
| Synthetic peptides corresponding to TMEM82(transmembrane protein 82) The peptide sequence was selected from the N terminal of TMEM82. Peptide sequence LETVHLAGLALFLTVVGSRVAALVVLEFSLRAVSTLLSLGKGSQGAAERL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| transmembrane protein 82 | |
| TMEM82 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title