missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM79 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563087
This item is not returnable.
View return policy
Description
Immunogen sequence: TLALLEVKRS DSPEKSSPQA LVPNGRQPEG EGGAESPGAE SLRVGSSAGS PTAIEGAEDG LDSTVSEAAT Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 70%.
TMEM79 is a multi-pass membrane protein. The exact function of TMEM79 remains unknown.
Specifications
| TMEM79 | |
| Polyclonal | |
| Unconjugated | |
| TMEM79 | |
| 2310042N02Rik; 2310074C17Rik; hypothetical protein MGC13102; MATT; Mattrin; RGD1309886; Tmem79; Transmembrane protein 79 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BSE2 | |
| TMEM79 | |
| Recombinant protein corresponding to Human TMEM79. Recombinant protein control fragment (Product #RP-101414). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction