missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM72 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49377-25ul
This item is not returnable.
View return policy
Description
TMEM72 Polyclonal antibody specifically detects TMEM72 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMEM72 | |
| Polyclonal | |
| Western Blot -Validated from a verified customer review, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| bA285G1.3, C10orf127, DKFZp686M0585, kidney-specific secretory protein of 37 kDa, KSP37, MGC157906, transmembrane protein 72 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LQPPNTLMELSLEPADSLAKKKQVHFEDNLVRIVPSLAEGLDD | |
| 25 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 643236 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction