missing translation for 'onlineSavingsMsg'
Learn More

TMEM70 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12849062 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849062 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849062 Supplier Thermo Scientific Supplier No. PA520915

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TMEM70 Polyclonal Antibody

TMEM70 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM70
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM70
Gene Accession No. Q921N7, Q9BUB7
Gene Alias 1110020A09Rik; 2210416J16Rik; MC5DN2; RGD1566224; TMEM70; transmembrane protein 70; transmembrane protein 70, mitochondrial
Gene Symbols TMEM70
Host Species Rabbit
Immunogen A 14 amino acid peptide near the center of human TMEM70.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500384, 54968, 70397
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.