missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM65 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98148AF750
This item is not returnable.
View return policy
Description
TMEM65 Polyclonal antibody specifically detects TMEM65 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TMEM65 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 157378 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| transmembrane protein 65 | |
| Produced in rabbits immunized with E. coli-derived Human TMEM65 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction