missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM63B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541883
This item is not returnable.
View return policy
Description
Peptide sequence: VRGCEQVEAI EYYTKLEQKL KEDYKREKEK VNEKPLGMAF VTFHNETITA Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
TMEM63B belongs to the SPO75/TMEM63 family. It is a multi-pass membrane protein. The function of TMEM63B remains unknown.
Specifications
| TMEM63B | |
| Polyclonal | |
| Unconjugated | |
| TMEM63B | |
| BC026370; C6orf110; CSC1-like protein 2; RGD1305862; RP3-421H19.2; Tmem63b; Tmem63b-ps1; Transmembrane protein 63B; transmembrane protein 63B, pseudogene 1; transmembrane protein 63B; CSC1-like protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55362 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5T3F8 | |
| TMEM63B | |
| Synthetic peptide directed towards the middle region of human TMEM63B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction