missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SLLDLFSTLC NDLVNSAQGF VSSTWTYYLQ TDNGKVVVFQ TQPIVESLGF QGGRLQRVEV TWRGSHPEAL EVHVDPVGPL DKVRKAKIRV KTSSKAKVES EEPQDNDFLS CMSR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
This gene encodes a predicted type-I membrane glycoprotein. The encoded protein may play a role in functioning of the central nervous system.
Specifications
Specifications
| Antigen | TMEM59L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM59L |
| Gene Accession No. | Q9UK28 |
| Gene Alias | 5330410G16Rik; brain-specific membrane-anchored protein; BSMAP; C19orf4; RGD1305557; Tmem59l; transmembrane protein 59 like; transmembrane protein 59-like |
| Gene Symbols | TMEM59L |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
