missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM59L Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81937CL1
This item is not returnable.
View return policy
Description
TMEM59L Polyclonal antibody specifically detects TMEM59L in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| TMEM59L | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| BSMAPBrain-specific membrane-anchored protein, C19orf4, chromosome 19 open reading frame 4, transmembrane protein 59-like | |
| TMEM59L antibody was raised against a 19 amino acid synthetic peptide near carboxy terminus of human TMEM59L. The immunogen is located within amino acids 220 - 270 of TMEM59L. Amino Acid Squence: RVKTSSKAKVESEEPQDND | |
| 0.1 mL | |
| Neuroscience | |
| 25789 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction