missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM59 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76497H
This item is not returnable.
View return policy
Description
TMEM59 Polyclonal antibody specifically detects TMEM59 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMEM59 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| C1orf8, chromosome 1 open reading frame 8, FLJ42977, HSPC001, Liver membrane-bound protein, transmembrane protein 59 | |
| Antibody was raised against a 16 amino acid synthetic peptide near amino terminus of human TMEM59. The immunogen is located within amino acids 80 - 130 of TMEM59. Amino Acid Squence: ECESACTEAYSQSDEQ | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction | |
| 9528 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction