missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM55B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561760
This item is not returnable.
View return policy
Description
Immunogen sequence: GHPAVLPGED PPPYSPLTSP DSGSAPMITC RVCQS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Catalyzes the hydrolysis of the 4-position phosphate of phosphatidylinositol 4,5-bisphosphate. Does not hydrolyze phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 3,4-bisphosphate, inositol 3,5-bisphosphate, inositol 3,4-bisphosphate, phosphatidylinositol 5-monophosphate, phosphatidylinositol 4-monophosphate and phosphatidylinositol 3-monophosphate.
Specifications
| TMEM55B | |
| Polyclonal | |
| Unconjugated | |
| PIP4P1 | |
| BC039161; C14orf9; PIP4P1; PtdIns-4,5-P(2) 4-phosphatase type I; ptdIns-4,5-P2 4-Ptase I; Tmem55b; Transmembrane protein 55B; type 1 phosphatidylinositol 4,5-bisphosphate 4-phosphatase; Type 1 PtdIns-4,5-P2 4-Ptase; type I phosphatidylinositol 4,5-bisphosphate 4-phosphatase; type I phosphatidylinositol-4,5-bisphosphate 4-phosphatase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90809 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86T03 | |
| PIP4P1 | |
| Recombinant protein corresponding to Human TMEM55B. Recombinant protein control fragment (Product #RP-99879). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction