Learn More
Invitrogen™ TMEM52B Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5144059
Product Code. 17901151
  • £362.00 / 100µg
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, monkey COS-7 whole cell, human SiHa whole cell, rat kidney tissue, rat NRK whole cell, mouse kidney tissue, mouse HBZY-1 whole cell. Flow: RT4 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The TMEM52B gene encodes a transmembrane protein that is widely expressed across various normal human tissues, suggesting its fundamental role in maintaining cellular integrity. TMEM52B is part of the transmembrane protein (TMEM) family, which contains proteins that span biological membranes, including the lipid bilayer of the plasma membrane and organelle membranes. This gene is notably involved in tumor biology, where it modulates cancer cell survival and invasion. Studies have shown that suppression of TMEM52B expression can enhance the aggressiveness of cancer cells, indicating its potential as a tumor suppressor gene. Specifically, TMEM52B isoforms such as P18 and P20 have been found to differentially influence tumor dynamics, particularly in the context of nasopharyngeal carcinoma (NPC). While its exact molecular mechanisms remain largely unknown, the regulation of TMEM52B is crucial for controlling tumor growth and metastasis. Therefore, understanding its role could provide insights into therapeutic strategies aimed at curbing cancer progression.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEM52B
Polyclonal
Unconjugated
TMEM52B
C12orf59; TMEM52B; Transmembrane protein 52B; UNQ5927/PRO19821
Rabbit
Antigen affinity chromatography
RUO
120939, 330428, 689963
-20°C
Lyophilized
ELISA, Flow Cytometry, Western Blot
500 μg/mL
PBS with 4mg trehalose and no preservative
Q0VBF2, Q4KMG9
TMEM52B
E.coli-derived human TMEM52B recombinant protein (Position: T22-N183).
100 μg
Primary
Human, Mouse, Monkey, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.