missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM51 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15967754
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15967754 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15967754 Supplier Invitrogen™ Supplier No. PA564979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EDSQEEEEEDE EAASRYYVPS YEEVMNTNYS EARGEEQNPR LSISLPSYES LTGLDETTPT STRADVEASP GNPPDRQNSK LAKRLKPLKV RRIKSEKLHL KDFRITLP Highest antigen sequence identity to the following orthologs - mouse 85%, rat 86%.

TMEM51 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM51
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM51
Gene Accession No. Q9NW97
Gene Alias C1orf72; TMEM51; Transmembrane protein 51
Gene Symbols TMEM51
Host Species Rabbit
Immunogen Recombinant protein fragment of Human TMEM51 (Product #RP-91047)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.