missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM50B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567028
This item is not returnable.
View return policy
Description
Immunogen sequence: DAAVVYPKPEQ LNHAFHTCG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
TMEM50B gene ontology annotations related to this gene include late endosome to vacuole transport via multivesicular body sorting pathway.
Specifications
| TMEM50B | |
| Polyclonal | |
| Unconjugated | |
| TMEM50B | |
| AU015466; AU019872; B230114J08Rik; C21orf4; HCV p7-transregulated protein 3; HCV p7-trans-regulated protein 3; HCVP7TP3; RGD1307561; TMEM50B; Transmembrane protein 50B; transmembrane protein 50B-like protein; UNQ167/PRO193 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 757 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P56557 | |
| TMEM50B | |
| Recombinant Human TMEM50B. Recombinant protein control fragment (Product #RP-107667). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction