missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566657
This item is not returnable.
View return policy
Description
Immunogen sequence: SEEWNPWEGDE KNEQQHRFKT SLQILDKSTK GKTDLSVQIW GKAAIGLYLW EHIFEGLLDP SDVTAQWREG KSIVGRTQ Highest antigen sequence identity to the following orthologs - mouse 82%, rat 81%.
Involved in the biosynthesis of the phosphorylated O-mannosyl trisaccharide (N-acetylgalactosamine-beta-3-N-acetylglucosamine-beta-4-(phosphate-6-)mannose), a carbohydrate structure present in alpha-dystroglycan (DAG1), which is required for binding laminin G-like domain-containing extracellular proteins with high affinity.
Specifications
| TMEM5 | |
| Polyclonal | |
| Unconjugated | |
| RXYLT1 | |
| 6330415D21Rik; HP10481; MDDGA10; Ribitol-5-phosphate xylosyltransferase 1; RXYLT1; TMEM5; Transmembrane protein 5; type II membrane protein; UDP-D-xylose:ribitol-5-phosphate beta1,4-xylosyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10329 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2B1 | |
| RXYLT1 | |
| Recombinant Human TMEM5. Recombinant protein control fragment (Product #RP-105298). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction