missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM48 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542844
This item is not returnable.
View return policy
Description
Peptide sequence: SFTEDRFGVV QTTLPAILNT LLTLQEAVDK YFKLPHASSK PPRISGSLVD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
TMEM48 is a component of the nuclear pore complex (NPC), which plays a key role in de novo assembly and insertion of NPC in the nuclear envelope. TMEM48 is required for NPC and nuclear envelope assembly, possibly by forming a link between the nuclear envelope membrane and soluble nucleoporins, thereby anchoring the NPC in the membrane.
Specifications
| TMEM48 | |
| Polyclonal | |
| Unconjugated | |
| NDC1 | |
| 2810475A17Rik; AI450313; FLJ10407; FLJ12556; FLJ34120; hNDC1; ndc1; NDC1 transmembrane nucleoporin; NET3; nuclear division cycle 1 homolog; nucleoporin NDC1; RP4-654H19.1; tmem48; Transmembrane protein 48; wu:fk93f04; zgc:55636 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55706 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BTX1 | |
| NDC1 | |
| Synthetic peptide directed towards the middle region of human TMEM48. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction