missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TMEM41B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL |
| Formulation | Tris Glycine |
| Gene Alias | KIAA0033MGC33897, MGC57262, transmembrane protein 41B |
| Host Species | Rabbit |
| Immunogen | Partial synthetic peptide made to an internal portion of the human TMEM41B protein (between amino acids 50-100) [UniProt Q5BJD5]. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?