missing translation for 'onlineSavingsMsg'
Learn More

TMEM41B Antibody, HRP, Novus Biologicals™

Product Code. 18355522 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355522 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355522 Supplier Novus Biologicals Supplier No. NBP312857H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM41B
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Formulation PBS without Preservative
Gene Alias KIAA0033MGC33897, MGC57262, transmembrane protein 41B
Host Species Rabbit
Immunogen Anti-TMEM41B antibody was raised against a peptide corresponding to 10 amino acids near the carboxyl terminus of human TMEM41B. The immunogen is located within the last 50 amino acids of TMEM41B.
Purification Method Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 440026
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.