missing translation for 'onlineSavingsMsg'
Learn More

TMEM41B Antibody, Biotin, Novus Biologicals™

Product Code. 18353634 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353634 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353634 Supplier Novus Biologicals Supplier No. NBP300527B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM41B Polyclonal specifically detects TMEM41B in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM41B
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias KIAA0033MGC33897, MGC57262, transmembrane protein 41B
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human TMEM41B protein (between amino acids 50-100) [UniProt Q5BJD5].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 440026
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.