missing translation for 'onlineSavingsMsg'
Learn More

TMEM38B Antibody, DyLight 405, Novus Biologicals™

Product Code. 18400505 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400505 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400505 Supplier Novus Biologicals Supplier No. NBP177093V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM38B Polyclonal antibody specifically detects TMEM38B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM38B
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias bA219P18.1, chromosome 9 open reading frame 87, D4Ertd89e, FLJ10493, transmembrane protein 38BC9orf87, TRIC-BTRICB, trimeric intracellular cation channel type B
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human TMEM38B. The immunogen is located within the last 50 amino acids of TMEM38B. Amino Acid Squence: VDVASDNVKKKHTKKNE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55151
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.