missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM38A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97578-100ul
This item is not returnable.
View return policy
Description
TMEM38A Polyclonal antibody specifically detects TMEM38A in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TMEM38A | |
| Polyclonal | |
| Western Blot 1:2000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| transmembrane protein 38ATRIC-AMGC3169, TRICA, trimeric intracellular cation channel type A | |
| Produced in rabbits immunized with E. coli-derived Human TMEM38A fragment. | |
| 100 μg | |
| Signal Transduction | |
| 79041 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction