missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567673
This item is not returnable.
View return policy
Description
TMEM37 Polyclonal Antibody for Western Blot
Thought to stabilize the calcium channel in an inactivated (closed) state. Modulates calcium current when coexpressed with CACNA1G.
Specifications
| TMEM37 | |
| Polyclonal | |
| Unconjugated | |
| TMEM37 | |
| AI173373; AW540203; Cacng5; Neuronal voltage-gated calcium channel gamma-like subunit; Pr; Pr1; protein distantly related to to the gamma subunit family; TMEM37; tmem37 {ECO:0000250; Transmembrane protein 37; UniProtKB:Q9JJV3}; voltage-dependent calcium channel gamma subunit-like protein; voltage-dependent calcium channel gamma-like subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140738 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WXS4 | |
| TMEM37 | |
| Synthesized peptide derived from human TMEM37. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction