Learn More
TMEM35 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24786-1-AP-20UL
 View more versions of this product
Product Code. 16803285
  • £135.00 / 20µL
Specifications
TMEM35
Polyclonal
Unconjugated
TMEM35A
TMEM35, transmembrane protein 35
Rabbit
Antigen Affinity Chromatography
RUO
59353, 67564
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q53FP2, Q9D328
TMEM35A
TMEM35 Fusion Protein Ag20589
20 μL
Primary
Mouse, Human