missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533080
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Opossum, Chicken (100%).
Acts as a regulator of the tubular endoplasmic reticulum (ER) network. Suppresses the RTN3/4-induced formation of the ER tubules. Positively regulates PERK-mediated and IRE1-mediated unfolded protein response signaling.
Specifications
| TMEM33 | |
| Polyclonal | |
| Unconjugated | |
| TMEM33 | |
| 1110006G02Rik; 1600019D15Rik; 2410089A21Rik; 2700052H22Rik; 5430406L04Rik; AA388285; AI314185; AI426615; db83; hypothetical protein LOC327214; Protein DB83; SHINC3; SHINC-3; Tmem33; Transmembrane protein 33; wu:fd16h10; wu:fi11f08; zgc:55931; zgc:77670 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 422779, 55161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P57088 | |
| TMEM33 | |
| Synthetic 18 amino acid peptide from internal region of human TMEM33. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Chicken | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction