missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121543
This item is not returnable.
View return policy
Description
TMEM27 Polyclonal Antibody for Western Blot, IHC (P)
TMEM27 (Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family. It is a stimulator of beta cell replication and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker. Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kDa. TMEM27 is an N-glycosylated protein and forms dimers, with MW of 64-75 kDa.
Specifications
| TMEM27 | |
| Polyclonal | |
| Unconjugated | |
| TMEM27 | |
| 0610008J07Rik; Cltrn; Collectrin; kidney-specific membrane protein; NX17; NX-17; Tmem27; Transmembrane protein 27; UNQ679/PRO1312 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 57395 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9ESG3 | |
| TMEM27 | |
| Recombinant protein Transmembrane Protein 27. The antigen corresponds to amino acid range 15-141 of the target protein. | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction