missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM261 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553197
This item is not returnable.
View return policy
Description
Immunogen sequence: MGSRLSQPFE SYITAPPGTA AAPAKPAPPA TPGAPTSPAE HRLLKTCWSC Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 41%.
Required for the assembly of the mitochondrial NADH:ubiquinone oxidoreductase complex (complex I). Involved in the assembly of the distal region of complex I. [UniProt].
Specifications
| TMEM261 | |
| Polyclonal | |
| Unconjugated | |
| DMAC1 | |
| 1700027K24Rik; 3110001D03Rik; C9orf123; distal membrane arm assembly complex 1; Distal membrane-arm assembly complex protein 1; Dmac1; Tmem261; Transmembrane protein 261; transmembrane protein C9orf123; transmembrane protein C9orf123 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90871 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GE9 | |
| DMAC1 | |
| Recombinant protein corresponding to Human TMEM261. Recombinant protein control fragment (Product #RP-90965). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction