missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM258 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560772
This item is not returnable.
View return policy
Description
Immunogen sequence: MELEAMSRYT SPVNPAVFPH LTVVL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
| TMEM258 | |
| Polyclonal | |
| Unconjugated | |
| TMEM258 | |
| C11orf10; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit TMEM258; HSPC005; Oligosaccharyl transferase subunit TMEM258; TMEM258; Transmembrane protein 258; UPF0197 transmembrane protein C11orf10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 746 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P61165 | |
| TMEM258 | |
| Recombinant protein corresponding to Human TMEM258. Recombinant protein control fragment (Product #RP-97349). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction