missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565669
This item is not returnable.
View return policy
Description
Immunogen sequence: GGTSTFTVTAH RAQHELNCSL QDPRSGRSAN ASVILNVQFK PEIAQVGAKY QEAQGPGLLV VLFALVRANP PANVTWIDQD GPVTVNTSDF LVLDAQNYPW LTNHTVQLQL RSLAHNLSVV ATNDVGVTSA SLPAPGLLAT RVEVPLLG Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
TMEM25 (Transmembrane Protein 25) is a Protein Coding gene. An important paralog of this gene is SIGLEC1.
Specifications
| TMEM25 | |
| Polyclonal | |
| Unconjugated | |
| TMEM25 | |
| 0610039J01Rik; AI429491; Tmem25; Transmembrane protein 25; UNQ2531/PRO6030 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84866 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86YD3 | |
| TMEM25 | |
| Recombinant Human TMEM25. Recombinant protein control fragment (Product #RP-106344). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction