Learn More
Invitrogen™ TMEM25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589785
Description
Immunogen sequence: APGPSRHPSL ISSDSNNLKL NNVRLPRENM SLPSNLQLND LTPDSRAVKP ADRQMAQNNS RPELLDPEPG GLLTSQGFIR LPVLGYIYRV SSVSSDEIWL; Positive Samples: U-87MG, OVCAR3, LO2, Mouse brain, Mouse kidney, Rat brain, Rat kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein.
TMEM25 (Transmembrane Protein 25) is a Protein Coding gene. An important paralog of this gene is SIGLEC1.
Specifications
| TMEM25 | |
| Polyclonal | |
| Unconjugated | |
| TMEM25 | |
| 0610039J01Rik; AI429491; Tmem25; Transmembrane protein 25; UNQ2531/PRO6030 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 689172, 71687, 84866 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.09 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q86YD3, Q9DCF1 | |
| TMEM25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 223-322 of human TMEM25 (NP_0011375061). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.