missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM25 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
£309.00
Specifications
| Antigen | TMEM25 |
|---|---|
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
Description
TMEM25 Polyclonal antibody specifically detects TMEM25 in Human samples. It is validated for ELISASpecifications
| TMEM25 | |
| ELISA | |
| Janelia Fluor 549 | |
| Rabbit | |
| Cell Biology | |
| 0610039J01Rik, FLJ14399, transmembrane protein 25 | |
| Produced in rabbits immunized with purified, recombinant Human TMEM25 (Accession#: AAH51841.1; Met1-Pro224) | |
| Antigen and protein A Affinity-purified |
| ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| 84866 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title