missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM24 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145107

Product Code. 18076351

  • £512.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LIISGVSKVP IAQDELALSL GYAASLEASV QDDAGTSGGP SSPPSDPPAM SPGPLDALSS PTSVQEADET TRSDISERPS VDDIESET Antigen sequence identity: Mouse 94%; Rat 93%.

Lipid-binding protein that transports phosphatidylinositol, the precursor of phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2), from its site of synthesis in the endoplasmic reticulum to the cell membrane (PubMed:28209843). It thereby maintains the pool of cell membrane phosphoinositides, which are degraded during phospholipase C (PLC) signaling (PubMed:28209843). Plays a key role in the coordination of Ca(2+) and phosphoinositide signaling: localizes to sites of contact between the endoplasmic reticulum and the cell membrane, where it tethers the two bilayers (PubMed:28209843). In response to elevation of cytosolic Ca(2+), it is phosphorylated at its C-terminus and dissociates from the cell membrane, abolishing phosphatidylinositol transport to the cell membrane (PubMed:28209843). Positively regulates insulin secretion in response to glucose: phosphatidylinositol transfer to the cell membrane allows replenishment of PI(4,5)P2 pools and calcium channel opening, priming a new population of insulin granules (PubMed:28209843). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMEM24
Polyclonal
Unconjugated
C2CD2L
1300006O23Rik; C2 calcium-dependent domain containing 2-like; C2 domain-containing protein 2-like; C2 domain-containing protein 2-like; phospholipid transfer protein C2CD2L; c2cd2 like; C2cd2l; C2CD2-like; DLNB23; KIAA0285; Phospholipid transfer protein C2CD2L; tmem24; Transmembrane protein 24; zgc:153961
Rabbit
Affinity Chromatography
RUO
9854
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
IgG
Immunocytochemistry, Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14523
C2CD2L
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 534-621. Recombinant protein control fragment (Product #a href=.
100 μL
Primary
Human
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.