missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM237 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539021
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a tetraspanin protein that is thought to be involved in WNT signaling. Defects in this gene are a cause of Joubert syndrome-14. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2012].
Specifications
| TMEM237 | |
| Polyclonal | |
| Unconjugated | |
| TMEM237 | |
| ALS2CR4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 381259, 65062 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96Q45, Q3V0J1 | |
| TMEM237 | |
| A synthetic peptide derived from the internal region of human ALS2CR4 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction