missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM237 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106602
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ALS2CR4.
The protein encoded by this gene is a tetraspanin protein that is thought to be involved in WNT signaling. Defects in this gene are a cause of Joubert syndrome-14. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2012].
Specifications
| TMEM237 | |
| Polyclonal | |
| Unconjugated | |
| TMEM237 | |
| AI853305; Als2cr4; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 4; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 4 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 4 protein homolog; Gm972; JBTS14; Tmem237; Transmembrane protein 237 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 381259, 65062 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3V0J1, Q96Q45 | |
| TMEM237 | |
| A synthesized peptide derived from human TMEM237(Accession Q96Q45), corresponding to amino acid residues V165-R215. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction