missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM231 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23731-1-AP-20UL
This item is not returnable.
View return policy
Description
The function remains unknown.Specifications
| TMEM231 | |
| Polyclonal | |
| Unconjugated | |
| TMEM231 | |
| ALYE870, FLJ22167, hypothetical protein FLJ22167, PRO1886, TMEM231, Transmembrane protein 231 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79583 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H6L2 | |
| TMEM231 | |
| TMEM231 Fusion Protein Ag20669 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction