missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM230 Antibody (OTI2C8), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71847PCP
This item is not returnable.
View return policy
Description
TMEM230 Monoclonal specifically detects TMEM230 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| TMEM230 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| TMEM230 | |
| Full length recombinant protein of human C20orf30 (NP_054864) produced in HEK293T cells. | |
| 0.1 mL | |
| 29058.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2C8 | |
| Flow Cytometry | |
| C20orf30, chromosome 20 open reading frame 30, dJ1116H23.2.1, HSPC274, hypothetical protein LOC29058, transmembrane protein 230 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction