missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM222 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525054
This item is not returnable.
View return policy
Description
TMEM222 Polyclonal Antibody for Western Blot, IHC (P)
The function of this protein has not been specifically defined.
Specifications
| TMEM222 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8BVA2, Q9H0R3 | |
| TMEM222 | |
| KLH conjugated synthetic peptide between 107-135 amino acids from the central region of human TM222. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| TMEM222 | |
| 5730406H10Rik; AA409341; C1orf160; D4Ertd196e; tm222; Tmem222; Transmembrane protein 222 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 52174, 84065 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction