missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM222 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112757
This item is not returnable.
View return policy
Description
TMEM222 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The function of this protein has not been specifically defined.
Specifications
| TMEM222 | |
| Polyclonal | |
| Unconjugated | |
| TMEM222 | |
| 5730406H10Rik; AA409341; C1orf160; D4Ertd196e; tm222; Tmem222; Transmembrane protein 222 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 84065 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H0R3 | |
| TMEM222 | |
| Recombinant Human Transmembrane protein 222 protein (77-164AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction