missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM217 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583773
This item is not returnable.
View return policy
Description
Immunogen sequence: SQGNIISYKR RISTAEILHS RNKRLSISSG FSGSHLESQY FERQSFHTSI FTCLSPVPSS APSTCRYTID V Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 30%.
TMEM217 is a protein coding gene. Gene ontology (GO) annotation include fibrillar center; integral component of membrane; nucleolus.
Specifications
| TMEM217 | |
| Polyclonal | |
| Unconjugated | |
| TMEM217 | |
| C6orf128; dJ355M6.2; TMEM217; Transmembrane protein 217 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221468 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N7C4 | |
| TMEM217 | |
| Recombinant protein corresponding to Human TMEM217. Recombinant protein control fragment (Product #RP-108365). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction