Learn More
TMEM211 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564290
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AFSLISPAWF QTPTFSFGIL TYCSWPQGNS WNQSCVTFSS LEDIPDFAWK V Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.
TMEM211 Polyclonal antibody specifically detects TMEM211 in Human samples. It is validated for Immunohistochemistry.Specifications
TMEM211 | |
Polyclonal | |
Unconjugated | |
TMEM211 | |
bA9F11.1; TMEM211; Transmembrane protein 211 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ICI0 | |
TMEM211 | |
Recombinant protein corresponding to Human TMEM211 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only