missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM200A Monoclonal Antibody (4-B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532881
This item is not returnable.
View return policy
Description
TMEM200A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| TMEM200A | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q86VY9, Q8C817 | |
| TMEM200A | |
| Recombinant protein within Human Transmembrane protein 200A aa 59-359. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 4-B3 | |
| Unconjugated | |
| TMEM200A | |
| C030003D03Rik; HBE61; Kiaa1913; Tmem200a; Transmembrane protein 200A; TTMA; TTMC; two transmembrane C | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 114801, 77220 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction