missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569170
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 85%; Rabbit: 100%; Rat: 100%.
This gene encodes a type II transmembrane protein that belongs to the interferon-induced transmembrane (IFITM) protein superfamily. The encoded protein functions as a cell surface hyaluronidase that cleaves extracellular high molecular weight hyaluronan into intermediate size fragments before internalization and degradation in the lysosome. It also has an interferon-mediated antiviral function in humans through activation of the JAK STAT signaling pathway. The activation of this gene by transcription factor SOX4 in breast cancer cells has been shown to mediate the pathological effects of SOX4 on cancer progression. Naturally occurring mutations in this gene are associated with autosomal recessive non-syndromic hearing loss.
Specifications
| TMEM2 | |
| Polyclonal | |
| Unconjugated | |
| CEMIP2 | |
| basic helix-loop-helix domain containing, class B, 2; BHLHB2; cell migration inducing hyaluronidase 2; Cell migration-inducing hyaluronidase 2; Cell surface hyaluronidase; CEMIP2; Kiaa1412; Tmem2; transmembrane protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83921 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5FWI3 | |
| CEMIP2 | |
| synthetic peptide directed towards the middle region of human TMEM2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction