missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM199 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25489-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| TMEM199 | |
| Polyclonal | |
| Unconjugated | |
| TMEM199 | |
| C17orf32, TMEM199, transmembrane protein 199 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 147007, 195040 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5SYH2, Q8N511 | |
| Tmem199 | |
| TMEM199 Fusion Protein Ag22009 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction